missing translation for 'onlineSavingsMsg'
Learn More

Ammonia, ca. 7N solution in methanol, AcroSeal™, Thermo Scientific Chemicals

76.78 € - 173.87 €

Specifications

Chemical Name or Material Ammonia
Color Colorless to Yellow
Name Note 7N solution in methanol
CAS 7664-41-7, 67-56-1
Molecular Formula H3N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10653415
View Documents
Thermo Scientific Acros
428381000
100 mL AcroSeal™ Glass Bottle
76.78 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10431864
View Documents
Thermo Scientific Acros
428388000
800 mL AcroSeal™ Glass Bottle
173.87 €
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ammonia
7N solution in methanol
H3N
MFCD00011418
0.7700g/mL
ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
N
17.03
CHEBI:16134
AcroSeal™ Glass Bottle
14°C
Colorless to Yellow
7664-41-7, 67-56-1
NH3
6.0 to 7.5N
15, 488
QGZKDVFQNNGYKY-UHFFFAOYSA-N
azane
222
17.03
0.77
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Causes damage to organs.
Fatal if inhaled.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF exposed: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

Product Identifier
  • Ammonia, ca 7N solution in methanol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H370-Causes damage to organs.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.