Learn More
Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals
Ammonia, H3N, CAS Number-7664-41-7, 67-56-1, am-fol, ammonia, ammonia anhydrous, ammonia, anhydrous, ammonia gas, ammoniak, ammoniakgas, anhydrous ammonia, nitro-sil, spirit of hartshorn, 2.5L, CHEBI:16134, Colorless to Yellow, 6.0 to 7.5N, 0.7700g/mL, 05,15, 14 deg.C, 17.03, GHS Signal Word: Danger
Brand: Thermo Scientific Acros 133710250
35.85 EUR valid until 2024-12-31
Use promo code "21665" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonia | |
ca. 7N Solution in Methanol | |
H3N | |
6.0 to 7.5N | |
05,15 | |
15, 488 | |
QGZKDVFQNNGYKY-UHFFFAOYSA-N | |
N | |
17.03 | |
222 | |
17.03 | |
0.77 |
Colorless to Yellow | |
7664-41-7, 67-56-1 | |
NH3 | |
MFCD00011418 | |
0.7700g/mL | |
ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol | |
25 mL | |
azane | |
14°C | |
CHEBI:16134 | |
Glass Bottle | |
Liquid |
RUO – Research Use Only