missing translation for 'onlineSavingsMsg'
Learn More

Ammonia Dilute R2 solution EU Pharmacopoeia, Fisher Chemical™

Product Code. 12997153
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

CAS 2032-59-9
Packaging Bottle
Quantity 1000 mL
Molecular Formula C11H16N2O2
InChI Key IMIDOCRTMDIQIJ-UHFFFAOYSA-N
SMILES CC1=C(C=CC(=C1)OC(=O)NC)N(C)C
IUPAC Name [4-(dimethylamino)-3-methylphenyl] N-methylcarbamate
Molecular Weight (g/mol) 208.261
PubChem CID 16247
ChEBI CHEBI:2653
Grade EU Ph Ref: 1004703
Analyte or Component Names Ammonia, dilute R2
Show More Show Less
compliance-icons
Product Identifier
  • Ammonia,dilute R2. European Pharmacopoeia Ref: 1004703. (RP 011)
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.