missing translation for 'onlineSavingsMsg'
Learn More
Ammonia solution 1mol/L, Fisher Chemical™ Product Code.: 12990744
compliance-icons compliance-icons compliance-icons

Ammonia solution 1mol/L, Fisher Chemical™
BIOPHARMA

Product Code. 12990744
100 mL
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy

Product Code. 12990744

Brand: Fisher Chemical SOL 105 100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
CAS 7664-41-7
Quantity 100 mL
Molecular Formula H3N
MDL Number MFCD00011418
Concentration 1M (1N)
InChI Key QGZKDVFQNNGYKY-UHFFFAOYSA-N
SMILES N
IUPAC Name azane
Molecular Weight (g/mol) 17.03
PubChem CID 222
ChEBI CHEBI:16134
Concentration or Composition (by Analyte or Components) 1mol/L
Chemical Name or Material Ammonia solution
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1087930 Certificate TypeCertificate of Analysis Date26/03/2025 Catalog NumberSOL 105 100
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonia 10M Volumetric Solution (SOL 105)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonia solution 1mol/L, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.