missing translation for 'onlineSavingsMsg'
Learn More

Ammonia Solution 20-22%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™

Product Code. 11934131 Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 mL
500 mL
1 L
Unit Size:
1 L
250mL
500mL
This item is not returnable. View return policy

Product Code. 11934131

Brand: Fisher Chemical A4701

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ammonia Solution 20-22%, Optima™, for Ultra Trace Elemental Analysis

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
CAS 1336-21-6
Molecular Formula H5NO
MDL Number 66650
Vapor Pressure 500 hPa 50
Vapor Density 0.59
pH 13.6
Quantity 1 L
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Ammonium hydroxide solution 20-22%, Optima for Ultra traces analysis
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.