missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia Solution 20-22%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™
Brand: Fisher Chemical A470-500
441.71 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
This item is not returnable.
View return policy
Description
Ammonia Solution 20-22%, Optima™, for Ultra Trace Elemental Analysis

Specifications
Colorless | |
500 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ammonium hydroxide solution 20-22%, Optima for Ultra traces analysis
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ammonia Solution 20-22%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™ > 500mL
Spot an opportunity for improvement?Share a Content Correction