missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia Solution 20-22%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
:
500mL
Specifications
Specifications
| pH | 13.6 |
| Color | Colorless |
| CAS | 1336-21-6 |
| Molecular Formula | H5NO |
| MDL Number | 66650 |
| Vapor Density | 0.59 |
| Vapor Pressure | 500 hPa 50 |
| Physical Form | Liquid |
Sécurité et traitement
Identifiant produit
- Ammonium hydroxide solution, 20-22%
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu