missing translation for 'onlineSavingsMsg'
Learn More

Ammonia solution 28%, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 10096010
Click to view available options
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Exceeds ACS Specifications

Specifications

Boiling Point 32°C to 37.7°C (1013 hPa)
CAS 1336-21-6
Melting Point -60°C to -57.5°C
Assay Percent Range 28.0 to 30.0%
Molecular Formula H5NO
MDL Number MFCD00066650
Density 0.88 - 0.91 g/cm³ (20°C)
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Quantity 1 L
SMILES N.O
IUPAC Name ammonium hydroxide
Molecular Weight (g/mol) 35.05
Residue after Ignition max. 0.001%
Specific Gravity 0.896 - 0.902 (at 60°)
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.