missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia solution, 2.0 M in methanol, Honeywell Fluka™
Click to view available options
Quantity:
100 mL
800 mL
2 L
4 x 100 mL
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 100 mL |
Linear Formula | NH3 |
UN Number | UN1992 |
Flash Point | 6°C (42.8°F) |
Formula Weight | 17.03g/mol |
Concentration or Composition (by Analyte or Components) | 2.0M in Methanol |
Packaging | Glass Bottle |
Density | 0.787g/mL at 25°C |
Chemical Name or Material | Ammonia solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'