missing translation for 'onlineSavingsMsg'
Learn More
Ammonia Solution, 35%, Certified AR for Analysis, d=0.88, Fisher Chemical™ Product Code.: 10049602

Ammonia Solution, 35%, Certified AR for Analysis, d=0.88, Fisher Chemical™

Product Code. 10049602
5 L, HDPE plastic bottle
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
HDPE plastic bottle
Plastic Coated Glass bottle
Plastic container
This item is not returnable. View return policy

Product Code. 10049602

Brand: Fisher Chemical A/3280/21

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

H5NO, CAS Number-1336-21-6, aquammonia, ammonium hydroxide solution, ammonium hydroxide nh4 oh, ammonia water, aqua ammonia, ammonia, monohydrate, ammonium hydroxide, ammonia aqueous, household ammonia, ammonia, aqua, 2.5L, CHEBI:18219, Colorless, 35.05g/mol, VHUUQVKOLVNVRT-UHFFFAOYSA-N

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILES N.O

Specifications

pH 12
Color Colorless
Quantity 5 L
IUPAC Name amine hydrate
Formula Weight 35.05g/mol
Packaging HDPE plastic bottle
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonia solution 35%
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Ammonium hydroxide,Ammonia
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonia Solution, 35%, Certified AR for Analysis, d=0.88, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.