missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia Solution, Strong, N.F. - F.C.C., J.T. Baker™
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Specifications
Quantity | 500 mL |
IUPAC Name | ammonia |
Formula Weight | 17.03 |
Grade | FCC/NF |
Packaging | Glass Bottle |
Specific Gravity | 1L = 0.90kg |
Chemical Name or Material | Ammonia Solution, Strong |
Safety and Handling
Signal Word
- Danger
- ACUTE AQUATIC Acute 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H400-Very toxic to aquatic life.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'