Learn More
Ammonium acetate, ACS, 97.0% min, Thermo Scientific Chemicals
Description
Ammonium acetate is a reagent used in chromatographic analysis of various compounds such as oligos, proteins, and peptides. It is also employed with acetic acid to provide a buffer solution. Generally it behaves as a catalyst in many reactions like Knoevenagel, Borch. It is also employed as a nutrient additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | ∽114°C (decomposition) |
Physical Form | Crystalline |
Quantity | 500 g |
Assay Percent Range | 97.0% min |
Beilstein | 4186741 |
Flash Point | 136°C (277°F) |
Solubility Information | Soluble in cold water,methanol ,SO2and acetone. |
Formula Weight | 77.08 |
Grade | ACS |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.