missing translation for 'onlineSavingsMsg'
Learn More
Ammonium Acetate, for HPLC, Certified, Fisher Chemical Product Code.: 10598410

Ammonium Acetate, for HPLC, Certified, Fisher Chemical

Product Code. 10598410
250g, Plastic powder jar
Click to view available options
250g, Plastic powder jar
This item is not returnable. View return policy

Product Code. 10598410

Brand: Fisher Chemical A/3446/50

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Specifications

Melting Point 114°C
CAS 631-61-8
Color White
pH 7
Physical Form Solid
Quantity 250 g
Molecular Formula C2H7NO2
MDL Number 13066
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
SMILES [NH4+].CC([O-])=O
IUPAC Name ammonium acetate
Molecular Weight (g/mol) 77.08
Formula Weight 77.08g/mol
Packaging Plastic powder jar
Show More Show Less
Product Identifier
  • Acetic acid, ammonium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium Acetate, for HPLC, Certified, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.