missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Acetate, LC-MS Ultra, Honeywell™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
Description
- For the most demanding LC-MS and UHPLC applications
- Tested for UHPLC-MS suitability
- Highest solvent purity for low detection limits
- Extremely low MS baseline noise.
- Exemplary lot-to-lot reproducibility
- Low levels of trace metals
- Packaged in plastic bottles

Specifications
Specifications
CAS | 631-61-8 |
Quantity | 25 g |
Molecular Formula | C2H7NO2 |
Linear Formula | CH3CO2NH4 |
MDL Number | MFCD00013066 |
Beilstein | 4186741 |
Synonym | ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 |
InChI Key | USFZMSVCRYTOJT-UHFFFAOYSA-N |
SMILES | CC(=O)[O-].[NH4+] |
IUPAC Name | azanium;acetate |
Show More |
Safety and Handling
Product Identifier
- Ammonium acetate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ammonium Acetate, LC-MS Ultra, Honeywell™ >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur