missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium bicarbonate, 99%, for analysis, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 105.0°C |
Color | White |
Physical Form | Crystalline Powder |
Quantity | 5 kg |
Assay Percent Range | 99% |
Linear Formula | NH4HCO3 |
Merck Index | 15, 494 |
Solubility Information | Solubility in water: 220g/L (20°C) |
Formula Weight | 79.06 |
Percent Purity | 99% |
Show More |
Safety and Handling
Product Identifier
- Ammonium bicarbonate
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'