missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium carbonate, ACS, NH{3} 30.0+%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
2 kg
Description
In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue, and fire extinguishing compoundsAmmonium carbonate is widely utilized as a leavening agent in lebkuchen, cookies and flat biscuits. It finds an important application as an emetic and an active ingredient
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue, and fire extinguishing compounds
Specifications
Specifications
Melting Point | 60°C (sublimation) |
Color | White |
Physical Form | Powder |
Quantity | 2 kg |
Assay Percent Range | NH3 30.0+% |
Molecular Formula | CH8N2O3 |
Beilstein | 3627235 |
Merck Index | 14,508 |
Synonym | ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate |
Solubility Information | Slowly soluble in water (1:4 portion); Decomposes in hot water. |
Show More |
Safety and Handling
Product Identifier
- Ammonium carbonate
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Ammonium carbonate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'