Learn More
Ammonium carbonate, ACS, NH{3} 30.0+%, Thermo Scientific Chemicals
Description
In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue, and fire extinguishing compoundsAmmonium carbonate is widely utilized as a leavening agent in lebkuchen, cookies and flat biscuits. It finds an important application as an emetic and an active ingredient
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue, and fire extinguishing compounds

Specifications
Specifications
Melting Point | 60° (sublimation) |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | NH3 30.0+% |
Molecular Formula | C2H11N3O5 |
Beilstein | 3627235 |
Merck Index | 14,508 |
Synonym | ammonium carbonate nf, ugam, ammonium bicarbonate-ammonium carbamate, ammonium carbonate chips nf, ammonium bicarbonate, mixt. with ammonium carbamate, ammonium carbamate, mixt. with ammonium bicarbonate, monoammonium carbonate mixture with ammonium carbamate, carbamic acid, monoammonium salt, mixt. with ammonium carbonate, diazanium hydrogen carbonate carbamate |
Solubility Information | Slowly soluble in water (1:4 portion); Decomposes in hot water. |
InChI Key | YNSUIDWLZCFLML-UHFFFAOYSA-N |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
R05J011 | Certificate of Analysis | 20/02/2024 | 036229.A3,036229.36 |
U04J005 | Certificate of Analysis | 16/08/2023 | 036229.A3,036229.36 |
M30J034 | Certificate of Analysis | 03/02/2023 | 036229.A3,036229.36 |
Z23H001 | Certificate of Analysis | 12/04/2022 | 036229.A3,036229.36 |
M10I031 | Certificate of Analysis | 12/04/2022 | 036229.A3,036229.36 |
Sicherheit und Handhabung

- Ammonium carbonate
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Ammonium carbonate
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.