missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Carbonate, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 100 g |
Linear Formula | (NH4)2CO3 |
Beilstein | 4253487 |
Formula Weight | 96.09g/mol |
Percent Purity | ≥30% (NH3 basis) |
Grade | ACS Reagent |
Density | Vapor Density: 2.7 (vs air) |
Chemical Name or Material | Ammonium Carbonate |
Safety and Handling
Product Identifier
- Ammonium carbonate
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Ammonium hydrogencarbonate, Ammonium carbamate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'