missing translation for 'onlineSavingsMsg'
Learn More

Ammonium carbonate, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 10734581
Click to view available options
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Meets ACS Specifications

Specifications

Melting Point 58°C
CAS 506-87-6
Quantity 1 kg
Assay Percent Range min. 30.0%
Molecular Formula CH8N2O3
MDL Number MFCD00010890
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
SMILES N.N.OC(O)=O
IUPAC Name carbonic acid diamine
Molecular Weight (g/mol) 96.09
Density 1.5 g/cm³ (20°C)
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium Carbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ammonium carbonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'