missing translation for 'onlineSavingsMsg'
Learn More

Ammonium carbonate, extra pure, Thermo Scientific Chemicals

Product Code. 10085303
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 506-87-6
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.09
MDL Number MFCD00010890
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
Synonym ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
PubChem CID 517111
IUPAC Name diazanium;carbonate
SMILES C(=O)([O-])[O-].[NH4+].[NH4+]

Spezifikation

Melting Point 58.0°C
Color White
Physical Form Powder
Quantity 1 kg
Linear Formula (NH4)2CO3
Merck Index 15, 505
Solubility Information Solubility in water: soluble (320g/L at 20°C)
Formula Weight 96.09
Percent Purity ≥30.5% (NH3)
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Ammonium carbonate
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'