missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium carbonate, for HPLC, 30 to 33% NH3 basis, Thermo Scientific™
Click to view available options
Quantity:
250 g
1 kg
Specifications
Melting Point | 58.0°C |
Quantity | 1 kg |
Linear Formula | (NH4)2CO3 |
Merck Index | 15, 505 |
Solubility Information | Solubility aq. soln.: soluble (320 g/L (at 20°C).) |
Formula Weight | 96.09 |
Grade | HPLC |
Packaging | Glass bottle |
Chemical Name or Material | Ammonium carbonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'