missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Carbonate (Lumps/Certified ACS), Fisher BioReagents

Product Code. 12857142
500g, Amber Glass
Click to view available options
500g, Amber Glass
This item is not returnable. View return policy
This item is not returnable. View return policy

This product is a mixture of Ammonium bicarbonate and Ammonium carbonate

Ammonium Carbonate is used in many biochemical procedures.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 58°C
CAS 8000-73-5
Color White
pH 9.4
Physical Form Solid
Quantity 500 g
Molecular Formula C2H11N3O5
Synonym ammonium carbonate nf, ugam, ammonium bicarbonate-ammonium carbamate, ammonium carbonate chips nf, ammonium bicarbonate, mixt. with ammonium carbamate, ammonium carbamate, mixt. with ammonium bicarbonate, monoammonium carbonate mixture with ammonium carbamate, carbamic acid, monoammonium salt, mixt. with ammonium carbonate, diazanium hydrogen carbonate carbamate
InChI Key YNSUIDWLZCFLML-UHFFFAOYSA-N
SMILES C(=O)(N)[O-].C(=O)(O)[O-].[NH4+].[NH4+]
IUPAC Name diazanium;hydrogen carbonate;carbamate
Molecular Weight (g/mol) 157.126
PubChem CID 24660
Percent Purity ≥30%
Grade Certified ACS
Identification Pass Test
Packaging Amber Glass
Vapor Pressure 69 hPa at 20°C
Chemical Name or Material Ammonium Carbonate
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium carbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium Carbonate (Lumps/Certified ACS), Fisher BioReagents >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.