missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium cerium(IV) nitrate solution, Honeywell™
177.64 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 16774-21-3 |
Quantity | 1 L |
Linear Formula | Ce(NH4)2(NO3)6 |
MDL Number | MFCD00151121 |
UN Number | UN2796 |
Formula Weight | 548.22g/mol |
Density | 1.10g/mL at 20°C |
Chemical Name or Material | Ammonium cerium(IV) nitrate solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'