missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium chloride, 99+%, pure, Thermo Scientific Chemicals
Ammonium chloride, 99+%, H4ClN, CAS Number-12125-02-9, amchlor, ammoneric, ammoniumchlorid, ammonium chloride, ammonium chloride nh4 cl, ammoniumklorid, ammonium muriate, darammon, sal ammoniac, salmiac, 1kg, 99% min. (Argentometry), CHEBI:31206, White, 1.5200g/mL, 235-186-4, 15,13, 53.49, Pure
Chemical Identifiers
CAS | 12125-02-9 |
---|---|
Molecular Formula | ClH4N |
Molecular Weight (g/mol) | 53.49 |
MDL Number | MFCD00011420 |
InChI Key | NLXLAEXVIDQMFP-UHFFFAOYSA-N |
Synonym | ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid |
PubChem CID | 25517 |
ChEBI | CHEBI:31206 |
SMILES | N.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10215410
|
Thermo Scientific Acros
123340250 |
25 g | Plastic Bottle |
N/A
|
N/A | ||||
10255360
|
Thermo Scientific Acros
123340010 |
1 kg | Plastic Bottle |
N/A
|
N/A | ||||
10266873
|
Thermo Scientific Acros
123340050 |
5 kg | Plastic drum |
N/A
|
N/A | ||||
10276873
|
Thermo Scientific Acros
123340251 |
25 kg | Fibre drum |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12125-02-9 | |
53.49 | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
25517 | |
N.Cl |
ClH4N | |
MFCD00011420 | |
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
CHEBI:31206 |
Specifications
340.0°C | |
White | |
99% min. (Argentometry) | |
NH3·HCl | |
15,13 | |
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
amine hydrochloride | |
25517 | |
53.49 | |
Pure | |
Ammonium chloride |
12125-02-9 | |
Crystalline Powder and/or Chunks | |
ClH4N | |
MFCD00011420 | |
15, 506 | |
Solubility in water: 370g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol,practically insoluble in acetone,ether and,ethyl acetate | |
N.Cl | |
53.49 | |
CHEBI:31206 | |
≥99% | |
1.5200g/mL |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not eat,d
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 235-186-4
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ammonium chloride
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only