missing translation for 'onlineSavingsMsg'
Learn More

Ammonium chloride, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5%

42.65 € - 817.51 €

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
View More Specs

Products 7
Produktcode Marke Quantity Preis Menge & Verfügbarkeit  
Produktcode Marke Quantity Preis Menge & Verfügbarkeit  
15673220
missing translation for 'viewDocuments'
Honeywell-Fluka
31107-500G
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15643220
missing translation for 'viewDocuments'
Honeywell-Fluka
31107-1KG
1 kg
73.79 €
1kg
Please to purchase this item. Need a web account? Register with us today!
15653220
missing translation for 'viewDocuments'
Honeywell-Fluka
31107-2.5KG
2.5 kg
157.97 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
15603230
missing translation for 'viewDocuments'
Honeywell-Fluka
31107-6X500G
6 x 500 g
241.54 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15683220
missing translation for 'viewDocuments'
Honeywell-Fluka
31107-6X1KG
6 x 1 kg
361.88 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15693220
missing translation for 'viewDocuments'
Honeywell-Fluka
31107-6X2.5KG
6 x 2.5 kg
794.34 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15663220
missing translation for 'viewDocuments'
Honeywell-Fluka
31107-25KG
25 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Beschreibung

Beschreibung

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Spezifikation

Chemische Identifikatoren

12125-02-9
53.49
NLXLAEXVIDQMFP-UHFFFAOYSA-N
25517
ClH4N
MFCD00011420
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
CHEBI:31206

Spezifikation

12125-02-9
NH4Cl
4371014
NLXLAEXVIDQMFP-UHFFFAOYSA-N
azanium;chloride
25517
53.49g/mol
ACS/puriss. p.a.
Vapor Density: 1.9 (vs air)
ClH4N
MFCD00011420
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
N.Cl
53.49
CHEBI:31206
≥99.5%
1mmHg (160.4°C)
Ammonium Chloride, Reag. Ph. Eur. reag. ISO
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

P301 + P312 + P330-P305 + P351 + P338

H302-H319-H411

missing translation for 'einecsNumber' : 235-186-4

missing translation for 'rtecsNumber' : BP4550000

Produktidentifikator
  • Ammonium chloride

Signalwort
  • Warning

Gefahrenkategorie
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2

Gefahrenhinweis
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.

Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt