missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Chloride, Reagent Grade, ≥99.5%, Honeywell™
Reagent Grade,≥99.5%
29.51 € - 77.79 €
Chemical Identifiers
CAS | 12125-02-9 |
---|---|
Molecular Formula | ClH4N |
Molecular Weight (g/mol) | 53.49 |
MDL Number | MFCD00011420 |
InChI Key | NLXLAEXVIDQMFP-UHFFFAOYSA-N |
Synonym | ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid |
PubChem CID | 25517 |
ChEBI | CHEBI:31206 |
IUPAC Name | azanium;chloride |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15637640
|
Honeywell Research Chemicals
A4514-100G |
100 g | Poly Bottle | ||||||
15657640
|
Honeywell Research Chemicals
A4514-500G |
500 g | Poly Bottle | ||||||
15647640
|
Honeywell Research Chemicals
A4514-1KG |
1 kg | Poly Bottle | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
12125-02-9 | |
53.49 | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
25517 | |
azanium;chloride |
ClH4N | |
MFCD00011420 | |
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
CHEBI:31206 |
Specifications
338°C | |
Colorless | |
Solid | |
NH4Cl | |
4371014 | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
azanium;chloride | |
25517 | |
53.49g/mol | |
Odorless | |
1.536g/cm3 at 20°C; Vapor Density: 1.9 (vs air) |
12125-02-9 | |
4.6 to 6 (at 20°C) | |
ClH4N | |
MFCD00011420 | |
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
[NH4+].[Cl-] | |
53.49 | |
CHEBI:31206 | |
≥99.5% | |
1mmHg (160.4°C) | |
Ammonium chloride |
Safety and Handling
P305 + P351 + P338
missing translation for 'rtecsNumber' : 235-186-4