missing translation for 'onlineSavingsMsg'
Learn More
Ammonium Chloride solution EU Pharmacopoeia, Fisher Chemical™ Product Code.: 12927163
compliance-icons

Ammonium Chloride solution EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA

Product Code. 12927163
1000 mL
Click to view available options
Quantity:
1000 mL
This item is not returnable. View return policy

Product Code. 12927163

Brand: Fisher Chemical RP 014 1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Packaging Bottle
Quantity 1000 mL
Grade EUR Ph Ref: 1005301
Chemical Name or Material Ammonium Chloride Solution

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2504007 Certificate TypeCertificate of Analysis Date09/04/2025 Catalog NumberRP 014 1000
Lot Number2503004 Certificate TypeCertificate of Analysis Date25/03/2025 Catalog NumberRP 014 1000
compliance-icons
Product Identifier
  • Ammonium chloride solution. European Pharmacopoeia Ref: 1005301. (RP 014)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium Chloride solution EU Pharmacopoeia, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.