missing translation for 'onlineSavingsMsg'
Learn More

Ammonium citrate tribasic, 97+%, Thermo Scientific Chemicals

Product Code. 11478806
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g

Lithium nitrate is used in ceramics, pyrotechnics, salt baths, heat-exchange media, refrigeration systems, rocket propellant and in solar batteries. It acts as an oxidizing agent used in the production of red-colored fireworks and flares, as a catalyst to accelerate the breakdown of nitrogen oxides and as a medium to store heat received from sun for cooking. It is applied to concrete-pavement to withstand weathering effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Solubility
Soluble in water. Slightly soluble in alcohol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3458-72-8
Molecular Formula C6H20N3O7+3
Molecular Weight (g/mol) 246.24
MDL Number MFCD00036406
InChI Key YWYZEGXAUVWDED-UHFFFAOYSA-Q
Synonym ammonium citrate
PubChem CID 131675891
IUPAC Name azane;[3-carboxy-1,3-dihydroxy-4-[oxonio(oxoniumylidene)methyl]butylidene]oxidanium
SMILES C(C(=[OH+])[OH2+])C(CC(=[OH+])O)(C(=O)O)O.N.N.N

Specifications

Melting Point ∼180°C (decomposition)
Assay Percent Range >97%
Quantity 100 g
Solubility Information Soluble in water. Slightly soluble in alcohol.
Formula Weight 243.22
Chemical Name or Material Ammonium citrate tribasic
compliance-icons
Product Identifier
  • Ammonium citrate tribasic
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Ammonium citrate tribasic

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.