missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium dichromate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Specifications
Melting Point | 170.0°C |
Color | Orange |
Physical Form | Adhering Crystals |
Quantity | 2.5 kg |
Assay Percent Range | 98.5% min. (Iodometry) |
Linear Formula | (NH4)2Cr2O7 |
Merck Index | 15, 512 |
Solubility Information | Solubility in water: 360g/L (20°C). Other solubilities: soluble in alcohol |
Formula Weight | 252.07 |
Percent Purity | 99% |
Show More |
Safety and Handling
Product Identifier
- Ammonium dichromate
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 1B
- Oxidising solid Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
- H272-May intensify fire; oxidiser.
- H301-Toxic if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360FD-May damage fertility. May damage the unborn child.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : Ammonium bichromate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction