Learn More
Ammonium Formate in Water, 10mM, with 0.05% Formic Acid, LC-MS , Thermo Scientific™
Mobile phase blend suitable for analysis such as vitamin D and drugs of abuse testing, and packaged in a borosilicate glass bottle that can be used directly on the LC-MS instrument.
Brand: Fisher Chemical MB123-1
Description
Ammonium Formate in Water, 10mM, with 0.05% Formic Acid, 0.57 to 0.69g/L (Ammonium Formate, w/v), CAS Number-7732-18-5, 540-69-2, 64-18-6, dihydrogen oxide, dihydrogen monoxide, 1L, <-0.005 AU at 254nm, <-1.0 AU at 220nm, Pass Test (LC-UV Gradient Suitability), <-5ppb, 100 deg.C, <-25ppb
- Suitable for use in LC/MS and UHPLC applications
- Pre-mixed for ease-of-use
- Mobile phase blend suitable for vitamin D and drugs of abuse testing
- Packaged in borosilicate glass bottles to prevent metal adduct formation, which can be used directly on the instrument.
- Ammonium formate, a commonly used HPLC buffer that is compatible with LC/MS
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
100°C | |
Mobile-phase solvent blend | |
3.5 | |
1 L | |
UHPLC-MS | |
Borosilicate Glass Bottle |
Ammonium Formate in Water, 10mM, with 0.05% Formic Acid | |
7732-18-5, 540-69-2, 64-18-6 | |
Liquid | |
dihydrogen oxide, dihydrogen monoxide | |
10mM Ammonium Formate | |
1.0 g/cm3 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only