missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Formate, purum p.a., Honeywell™

Product Code. 15640900
Click to view available options
Quantity:
100 g
500 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Purum p.a.,≥97.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-69-2
Molecular Formula CH5NO2
Molecular Weight (g/mol) 63.056
MDL Number MFCD00013103
InChI Key VZTDIZULWFCMLS-UHFFFAOYSA-N
Synonym ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny
PubChem CID 2723923
ChEBI CHEBI:63050
IUPAC Name azanium;formate
SMILES C(=O)[O-].[NH4+]

Specifications

Quantity 1 kg
Linear Formula HCO2NH4
UN Number NONH for all modes of transport
Beilstein 3625095
Formula Weight 63.06g/mol
Percent Purity ≥97% (T)
Grade purum p.a.
Density 1.26g/mL at 25°C
Chemical Name or Material Ammonium Formate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'