missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Formate, purum p.a., Honeywell™
Purum p.a.,≥97.0% (T)
23.84 € - 82.18 €
Chemical Identifiers
CAS | 540-69-2 |
---|---|
Molecular Formula | CH5NO2 |
Molecular Weight (g/mol) | 63.056 |
MDL Number | MFCD00013103 |
InChI Key | VZTDIZULWFCMLS-UHFFFAOYSA-N |
Synonym | ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny |
PubChem CID | 2723923 |
ChEBI | CHEBI:63050 |
IUPAC Name | azanium;formate |
SMILES | C(=O)[O-].[NH4+] |
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
540-69-2 | |
63.056 | |
VZTDIZULWFCMLS-UHFFFAOYSA-N | |
2723923 | |
azanium;formate |
CH5NO2 | |
MFCD00013103 | |
ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny | |
CHEBI:63050 | |
C(=O)[O-].[NH4+] |
Specifications
540-69-2 | |
HCO2NH4 | |
NONH for all modes of transport | |
ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny | |
C(=O)[O-].[NH4+] | |
63.056 | |
CHEBI:63050 | |
≥97% (T) | |
1.26g/mL at 25°C |
CH5NO2 | |
MFCD00013103 | |
3625095 | |
VZTDIZULWFCMLS-UHFFFAOYSA-N | |
azanium;formate | |
2723923 | |
63.06g/mol | |
purum p.a. | |
Ammonium Formate |
Safety and Handling
missing translation for 'einecsNumber' : 208-753-9
missing translation for 'rtecsNumber' : BQ6650000