missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydrogen difluoride, 95%, extra pure, Thermo Scientific Chemicals

Product Code. 10296973
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1341-49-7
Molecular Formula F2H5N
Molecular Weight (g/mol) 57.04
MDL Number MFCD00012018
InChI Key KVBCYCWRDBDGBG-UHFFFAOYSA-N
Synonym Ammonium bifluoride
IUPAC Name amine dihydrofluoride
SMILES N.F.F

Specifications

Boiling Point 230.0°C
Melting Point 125.0°C
Color White
Physical Form Flakes or Scales
Quantity 250 g
Assay Percent Range 95%
Linear Formula (NH4)HF2
Merck Index 15, 495
Solubility Information Solubility aq. soln.: 630 g/L (20°C). Other solubilities: very soluble in hot water, soluble in ethanol
Formula Weight 57.04
Percent Purity 95%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Ammonium hydrogen difluoride, tech., 95%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'