missing translation for 'onlineSavingsMsg'
Learn More
Ammonium hydroxide, 0.1N Standardized Solution, Thermo Scientific Chemicals Product Code.: 11333677

Ammonium hydroxide, 0.1N Standardized Solution, Thermo Scientific Chemicals

Product Code. 11333677
4 L
Click to view available options
Quantity:
1 L
4 L
This item is not returnable. View return policy

Product Code. 11333677

Brand: Thermo Scientific Alfa Aesar 035576.K7

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 ohShow MoreShow Less
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name azanium;hydroxide
SMILES N.O

Specifications

Concentration 0.1N
Formula Weight 35.05
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Ammonium hydroxide, Standardized Solution

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM31L511 Certificate TypeCertificate of Analysis Date06/02/2025 Catalog Number035576.K7,035576.K2
Lot NumberZ14K524 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog Number035576.K7,035576.K2
Lot NumberW18K514 Certificate TypeCertificate of Analysis Date23/09/2024 Catalog Number035576.K7,035576.K2
Lot NumberZ01J526 Certificate TypeCertificate of Analysis Date18/01/2024 Catalog Number035576.K7,035576.K2
Lot NumberX06J537 Certificate TypeCertificate of Analysis Date16/11/2023 Catalog Number035576.K7,035576.K2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium hydroxide, 0.1N Standardized Solution, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.