missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium hydroxide, ACS reagent, 28-30% solution in water, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
10 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Colorless |
Boiling Point | 38.0°C |
Melting Point | -57.0°C |
Linear Formula | NH4OH |
Fieser | 05,15 |
Density | 0.9100g/mL |
Merck Index | 15, 491 |
Solubility Information | Solubility in water: soluble. |
Quantity | 10 L |
IUPAC Name | amine hydrate |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'