Learn More
Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water, Thermo Scientific Chemicals
35.05 € - 271.35 €
Chemical Identifiers
CAS | 1336-21-6, 7732-18-5 |
---|---|
Molecular Formula | H5NO |
Molecular Weight (g/mol) | 35.05 |
MDL Number | MFCD00066650 |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
Synonym | ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 ohShow More |
PubChem CID | 14923 |
ChEBI | CHEBI:18219 |
SMILES | N.O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10470481
|
Thermo Scientific Acros
205840010 |
1 L | Glass bottle |
35.05 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10105220
|
Thermo Scientific Acros
205840025 |
2.5 L | Glass bottle |
65.93 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10134423
|
Thermo Scientific
205840100 |
10 L | Plastic drum |
198.00 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15294674
|
Thermo Scientific Acros
205840250 |
25 L | Plastic Can |
271.35 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1336-21-6, 7732-18-5 | |
35.05 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
14923 | |
N.O |
H5NO | |
MFCD00066650 | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 ohShow More | |
CHEBI:18219 |
Specifications
38.0°C | |
-57.0°C | |
NH4OH | |
0.9100g/mL | |
Solubility in water: soluble. | |
N.O | |
35.05 | |
CHEBI:18219 | |
Analytical | |
Ammonium hydroxide |
1336-21-6, 7732-18-5 | |
H5NO | |
MFCD00066650 | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
amine hydrate | |
14923 | |
35.05 | |
0.91 |
Safety and Handling
GHS P Statement
May cause respiratory irritation.
Very toxic to aquatic life.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Ammonium hydroxide, 25-30% solution in water
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Ammonium hydroxide,Ammonia
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.