missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium hydroxide solution, Eluent additive for LC-MS, ≥25% in H2O, Honeywell Fluka™
Eluent additive for LC-MS,≥25% in H2O
71.85 € - 116.23 €
Chemical Identifiers
CAS | 1336-21-6, 7732-18-5 |
---|---|
Molecular Formula | H5NO |
Molecular Weight (g/mol) | 35.05 |
MDL Number | MFCD00066650 |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
Synonym | ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh |
PubChem CID | 14923 |
ChEBI | CHEBI:18219 |
SMILES | N.O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15628410
|
Honeywell-Fluka
44273-10X1ML |
10 x 1 mL | Ampoule | ||||||
Currently Unavailable: We have no information when this product will be available
|
15655540
|
Honeywell-Fluka
44273-100ML-GL |
100 mL | Glass Bottle | |||||
15645540
|
Honeywell-Fluka
44273-100ML |
100 mL | Polybottle |
116.23 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
1336-21-6, 7732-18-5 | |
35.05 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
14923 | |
N.O |
H5NO | |
MFCD00066650 | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
CHEBI:18219 |
Specifications
24°C | |
≥70.0000%, ≥20.0000% | |
H5NO | |
MFCD00066650 | |
3587154 | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
N.O | |
0.27 | |
14923 | |
35.05 | |
LC-MS |
1336-21-6, 7732-18-5 | |
<90.0000%, <30.0000% | |
NH4OH | |
UN2672 | |
0.9 g/mL at 25 °C(lit.) | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
azanium;hydroxide | |
35.05 | |
CHEBI:18219 | |
>25% | |
Ammonium hydroxide solution, Eluent additive for LC-MS, ≥25% in H2O |
Safety and Handling
P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P391
missing translation for 'shelfLife' : 1080 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Ammonia solutions
Recommended Storage : Room Temp
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Ammonium hydroxide solution
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Ammonia, aqueous solution