missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium hydroxide solution, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.

Specifications
Specifications
CAS | 1336-21-6 |
Molecular Formula | H5NO |
MDL Number | MFCD00066650 |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
Quantity | 2.5 L |
SMILES | N.O |
IUPAC Name | amine hydrate |
Molecular Weight (g/mol) | 35.05 |
PubChem CID | 14923 |
ChEBI | CHEBI:18219 |
Show More |
Safety and Handling



Product Identifier
- Ammonium hydroxide solution
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Ammonia
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ammonium hydroxide solution, Honeywell™ >
Spot an opportunity for improvement?Share a Content Correction