missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(II) sulfate hexahydrate, 99+%, for analysis

Product Code. 10762241 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Unit Size:
1kg
25g
25kg
500g
5kg
This item is not returnable. View return policy

Product Code. 10762241

Brand: Thermo Scientific Acros 201370250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-85-9 | FeH20N2O14S2 | 392.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-85-9
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
Synonym Ferrous ammonium sulfate, Mohr's salt
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point 100.0°C
Color Green
Physical Form Crystalline Powder and/or Crystals
Quantity 25 g
Assay Percent Range 99+%
Linear Formula (NH4)2Fe(SO4)2·6H2O
Merck Index 15, 518
Solubility Information Solubility in water: 269g/L (20°C). Other solubilities: practically insoluble in alcohol
Formula Weight 392.14
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Ammonium iron(II) sulfate hexahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium iron (II) sulfate hexahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis