missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Iron(III) Sulfate Dodecahydrate, Certified AR for analysis, Fisher Chemical™
134.65 EUR valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
Description
H4FeNO8S2.12H2O, CAS Number-7783-83-7, 500g, 230 deg.C, Purple, 482.19g/mol, 39 deg.C, 150004, Plastic powder jar, 2.5, Solid
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 230°C |
Melting Point | 39°C |
Color | Purple |
pH | 2.5 |
Physical Form | Solid |
Quantity | 1 kg |
Formula Weight | 482.19g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'