missing translation for 'onlineSavingsMsg'
Learn More

Ammonium metavanadate, 99.5%, for analysis, Thermo Scientific Chemicals

44.42 € - 1287.41 €

Chemical Identifiers

CAS 7803-55-6
Molecular Formula H4NO3V
Molecular Weight (g/mol) 116.98
MDL Number MFCD00011430
InChI Key YBVKNHXQSRDWAA-UHFFFAOYSA-M
Synonym ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
PubChem CID 516859
SMILES N.O[V](=O)=O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10246300
View Documents
Thermo Scientific Acros
194910500
50 g Glass bottle
44.42 €
50g
Please to purchase this item. Need a web account? Register with us today!
10695452
View Documents
Thermo Scientific Acros
194912500
250 g Glass bottle
115.58 €
250g
Please to purchase this item. Need a web account? Register with us today!
10185443
View Documents
Thermo Scientific Acros
194910010
1 kg Glass bottle
319.59 €
1kg
Please to purchase this item. Need a web account? Register with us today!
11984331
View Documents
Thermo Scientific Acros
194910050
5 kg Plastic Drum
1287.41 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7803-55-6
116.98
YBVKNHXQSRDWAA-UHFFFAOYSA-M
516859
H4NO3V
MFCD00011430
ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
N.O[V](=O)=O

Specifications

200.0°C
Green to White
99.5%
NH4VO3
01,1057
ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
YBVKNHXQSRDWAA-UHFFFAOYSA-M
azanium;oxido(dioxo)vanadium
516859
99.5%
Ammonium metavanadate
7803-55-6
Crystalline Powder
H4NO3V
MFCD00011430
15,565
Solubility in water: 5.1g/L (20°C). Other solubilities: insoluble in ethanol, 69.5g/L (96°C)
N.O[V](=O)=O
116.98
116.98
Analytical
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of damaging the unborn child.
Causes damage to organs through prolonged or repeated exposure if inhaled.
Toxic if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Toxic to aquati

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 232-261-3

Product Identifier
  • Ammonium metavanadate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1

Hazard Statement
  • H372-Causes damage to organs.
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.
  • H411-Toxic to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only