missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate (para) tetrahydrate, 99%

Product Code. 11494814
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11494814

Brand: Thermo Scientific Alfa Aesar A13766.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12054-85-2 | (NH4)6Mo7O24·4H2O

Ammonium molybdate tetrahydrate is widely utilized in cryo-negative staining which helps to visualize particles while preserving the sample. It is used as a catalyst to desulfurize thiophene and its derivatives. It serves as an analytical reagent to measure the amount of phosphates, silicates, arsenates and lead in an aqueous solution. It is also used in electroplating, in fertilizers for crops and in the production of molybdenum metal and ceramics. It is used as an analgesic in medical field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium molybdate tetrahydrate is widely utilized in cryo-negative staining which helps to visualize particles while preserving the sample. It is used as a catalyst to desulfurize thiophene and its derivatives. It serves as an analytical reagent to measure the amount of phosphates, silicates, arsenates and lead in an aqueous solution. It is also used in electroplating, in fertilizers for crops and in the production of molybdenum metal and ceramics. It is used as an analgesic in medical field.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12054-85-2
Molecular Formula (NH4)6Mo7O24·4H2O
MDL Number MFCD00167059
Synonym Ammonium heptamolybdate tetrahydrate; Ammonium paramolybdate tetrahydrate

Specifications

Melting Point ∼190°C (decomposition)
Quantity 250 g
Merck Index 14,533
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 1235.86 (1163.79 Anhydrous)
Percent Purity 99%
Density 2.498
Chemical Name or Material Ammonium molybdate (para) tetrahydrate
compliance-icons
Product Identifier
  • Ammonium molybdate (para) tetrahydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Ammonium heptamolybdate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.