Learn More
Ammonium molybdate (para) tetrahydrate, ACS, 81-83% as MoO{3}, Thermo Scientific Chemicals
Ammonium molybdate (para) tetrahydrate, H24Mo7N6O24.4H2O, CAS Number-12054-85-2, Ammonium paramolybdate, 2kg, Powder, 81-83%, 2.498, 234-722-4, 1235.86 (1163.79 Anhydrous), ACS, H302-H315-H319-H335, P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
45.80 € - 558.00 €
Chemical Identifiers
CAS | 12054-85-2 |
---|---|
Molecular Formula | H24Mo7N6O24·4H2O |
MDL Number | MFCD00167059 |
Synonym | Ammonium paramolybdate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11341878
|
Thermo Scientific Alfa Aesar
011831.18 |
50 g |
45.80 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11351878
|
Thermo Scientific Alfa Aesar
011831.22 |
100 g |
56.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11321878
|
Thermo Scientific Alfa Aesar
011831.36 |
500 g |
175.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11331878
|
Thermo Scientific Alfa Aesar
011831.A3 |
2 kg |
558.00 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ammonium molybdate (para) tetrahydrate, is used in photography,analytical lab reagent for phosphate,lead,arsenate determination.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12054-85-2 | |
H24Mo7N6O24·4H2O | |
MFCD00167059 | |
Ammonium paramolybdate |
Specifications
Powder | |
∽190°C (decomposition) | |
H24Mo7N6O24·4H2O | |
2.498 g/cm3 | |
Soluble in water (1:2 ratio). Insoluble in alcohol | |
ACS | |
Powder |
12054-85-2 | |
81-83% | |
MFCD00167059 | |
Ammonium paramolybdate | |
1235.86 (1163.79 Anhydrous) | |
Odorless | |
Ammonium molybdate (para) tetrahydrate, (para) tetrahydrate |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 234-722-4
missing translation for 'rtecsNumber' : QA4900000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only