missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate(VI) tetrahydrate, ACS reagent

Product Code. 10773223 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
5g
This item is not returnable. View return policy

Product Code. 10773223

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

CAS: 12054-85-2 | H24Mo7N6O24·4H2O | 1235.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12054-85-2
Molecular Formula H24Mo7N6O24·4H2O
Molecular Weight (g/mol) 1235.86
MDL Number MFCD00167059

Specifications

Melting Point 190°C
Color White
Physical Form Crystalline Powder
Quantity 100 g
Linear Formula (NH4)6Mo7O24·4H2O
Merck Index 15, 530
Solubility Information Solubility in water: 400g/L (20°C). Other solubilities: practically insoluble in alcohol
Formula Weight 1235.86
Percent Purity 81.0 to 83.0% (MoO3)
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 2.49
Decomposition Information >150°C
Density 2.4900g/mL
Chemical Name or Material Ammonium molybdate(VI) tetrahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium molybdate(VI) tetrahydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Ammonium heptamolybdate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.