missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate(VI) tetrahydrate, ACS reagent, Thermo Scientific Chemicals

24.52 € - 831.14 €

Chemical Identifiers

CAS 12054-85-2
Molecular Formula H24Mo7N6O24·4H2O
Molecular Weight (g/mol) 1235.86
MDL Number MFCD00167059
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10792402
View Documents
Thermo Scientific Acros
423310050
5 g Glass Bottle
24.52 €
5g
Please to purchase this item. Need a web account? Register with us today!
10773223
View Documents
Thermo Scientific Acros
423311000
100 g Plastic bottle
46.13 €
100g
Please to purchase this item. Need a web account? Register with us today!
10792592
View Documents
Thermo Scientific Acros
423315000
500 g Glass bottle
184.92 €
500g
Please to purchase this item. Need a web account? Register with us today!
10562714
View Documents
Thermo Scientific Acros
423310025
2.5 kg Plastic bottle
831.14 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

12054-85-2
H24Mo7N6O24·4H2O
1235.86
MFCD00167059

Specifications

190°C
White
H24Mo7N6O24·4H2O
MFCD00167059
Solubility in water: 400g/L (20°C). Other solubilities: practically insoluble in alcohol
1235.86
ACS Reagent
>150°C
Ammonium molybdate(VI) tetrahydrate
12054-85-2
Crystalline Powder
(NH4)6Mo7O24·4H2O
15, 530
1235.86
81.0 to 83.0% (MoO3)
2.49
2.4900g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only