Learn More
Ammonium nitrate, 99+%, for analysis, Thermo Scientific™
Ammonium nitrate, 99+%, H4N2O3, CAS Number-6484-52-2, ammonium nitricum, ammonium saltpeter, merco prills, herco prills, nitric acid ammonium salt, nitram, nitrato amonico, nitrate of ammonia, german saltpeter, ammonium nitrate, 500g, 210.0 deg.C, CHEBI:63038, 5ppm max, 5ppm max., White, 1.7200g/mL
Chemical Identifiers
| CAS | 6484-52-2 |
|---|---|
| Molecular Formula | H4N2O3 |
| Molecular Weight (g/mol) | 80.04 |
| MDL Number | MFCD00011425 |
| InChI Key | DVARTQFDIMZBAA-UHFFFAOYSA-O |
| Synonym | ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills |
| PubChem CID | 22985 |
| ChEBI | CHEBI:63038 |
| IUPAC Name | azanium;nitrate |
| SMILES | [NH4+].[N+](=O)([O-])[O-] |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
For analysisChemical Identifiers
| H4N2O3 | |
| MFCD00011425 | |
| ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills | |
| CHEBI:63038 | |
| [NH4+].[N+](=O)([O-])[O-] |
Specifications
| 6484-52-2 | |
| H4N2O3 | |
| MFCD00011425 | |
| 15,531 | |
| Solubility in water: 190g/100mL (20°C). Other solubilities: 1g/20mL alcohol,1g/8mL methanol | |
| [NH4+].[N+](=O)([O-])[O-] | |
| 80.04 | |
| CHEBI:63038 | |
| 99+% | |
| 0.01% max. (as SO4) | |
| 1.7200g/mL |
| 99+% | |
| NH4NO3 | |
| 01,39; 15,14 | |
| ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills | |
| DVARTQFDIMZBAA-UHFFFAOYSA-O | |
| azanium;nitrate | |
| 22985 | |
| 80.04 | |
| Analysis | |
| 1.72 | |
| Ammonium nitrate, p.a. |
Safety and Handling
GHS H Statement
May intensify fire; oxidizer.
Causes serious eye irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/ combustible materials.
In case of fire: Use water spray for extinction.
Wear protective gloves/protective clothing/
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 229-347-8
missing translation for 'rtecsNumber' : BR9050000
missing translation for 'tsca' : TSCA
- Ammonium nitrate
Signal Word
- Warning
Hazard Category
- Oxidising solid Category 3
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H272-May intensify fire; oxidiser.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P370+P378-In case of fire: Use to extinguish.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.