Learn More
Ammonium oxalate monohydrate, 98%
Description
Ammonium oxalate monohydrate is widely utilized as a buffering reagent and a dispersant to determine the surface interaction of platelets. It finds an application to study its acute poisoning effect on sheep and to investigate the formation of oxalate monoalkylamide in the human lens. It is also used in the detection and determination of calcium, lead, fluoride and rare earth metals. It is employed as chelators and forms complexes with metals. It acts as reducing agent in gold extraction and is an active ingredient of ferrous metal surface polishing fluid. It is a promoting agent in production of cobalt oxide and nickel oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ammonium oxalate monohydrate is widely utilized as a buffering reagent and a dispersant to determine the surface interaction of platelets. It finds an application to study its acute poisoning effect on sheep and to investigate the formation of oxalate monoalkylamide in the human lens. It is also used in the detection and determination of calcium, lead, fluoride and rare earth metals. It is employed as chelators and forms complexes with metals. It acts as reducing agent in gold extraction and is an active ingredient of ferrous metal surface polishing fluid. It is a promoting agent in production of cobalt oxide and nickel oxide.
Solubility
Soluble in water. Slightly soluble in alcohol.
Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications
Spezifikation
| Melting Point | 70°C (decomposition) |
| Density | 1.5 |
| Assay Percent Range | >99% |
| Linear Formula | (COONH4)2·H2O |
| Quantity | 500 g |
| UN Number | UN3288 |
| Beilstein | 3759830 |
| Merck Index | 14,537 |
| Solubility Information | Soluble in water. Slightly soluble in alcohol. |
| Formula Weight | 142.11 (124.10 Anhydrous) |
| Mehr anzeigen |
Sicherheit und Handhabung
- Ammonium oxalate monohydrate
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Ammonium oxalate monohydrate
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.