missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Perchlorate Solution, Honeywell

Chemical Identifiers

CAS 7790-98-9
Molecular Formula ClH4NO4
Molecular Weight (g/mol) 117.485
MDL Number MFCD00011421
InChI Key HHEFNVCDPLQQTP-UHFFFAOYSA-N
PubChem CID 24639
IUPAC Name azanium;perchlorate
SMILES [NH4+].[O-]Cl(=O)(=O)=O
View More Specs

Description

Description

Specifications

Chemical Identifiers

7790-98-9
117.485
HHEFNVCDPLQQTP-UHFFFAOYSA-N
azanium;perchlorate
ClH4NO4
MFCD00011421
24639
[NH4+].[O-]Cl(=O)(=O)=O

Specifications

7790-98-9
NH4ClO4
3211
[NH4+].[O-]Cl(=O)(=O)=O
117.485
117.49g/mol
Ammonium Perchlorate Solution
ClH4NO4
MFCD00011421
HHEFNVCDPLQQTP-UHFFFAOYSA-N
azanium;perchlorate
24639
Reagent
Videos
Safety and Handling

Safety and Handling

P220-P305 + P351 + P338

H272-H319-H373

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.