missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Persulfate, PlusOne™, Cytiva

Product Code. 10005184
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Supports efficient polymerization of acrylamide at concentrations as low as 1mM

TRUSTED_SUSTAINABILITY

Specifications

CAS 7727-54-0
Quantity 25 g
Assay Percent Range >98%
Molecular Formula H8N2O8S2
MDL Number MFCD00003390
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
IUPAC Name diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular Weight (g/mol) 228.19
PubChem CID 62648
Formula Weight 228.2
Chemical Name or Material Ammonium Persulfate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger||
Hazard Category
  • Oxidising solid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/incompatible materials/combustible materials.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ammonium peroxodisulphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.