Learn More
Ammonium phosphate, dibasic, 99+%, for analysis, Thermo Scientific Chemicals
35.18 € - 231.15 €
Chemical Identifiers
CAS | 7783-28-0 |
---|---|
Molecular Formula | H9N2O4P |
Molecular Weight (g/mol) | 132.06 |
MDL Number | MFCD00010891 |
InChI Key | MNNHAPBLZZVQHP-UHFFFAOYSA-N |
Synonym | diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate |
PubChem CID | 24540 |
ChEBI | CHEBI:63051 |
IUPAC Name | phosphoric acid diamine |
SMILES | N.N.OP(O)(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10609693
|
Thermo Scientific Acros
201822500 |
250 g | Plastic bottle |
35.18 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10123980
|
Thermo Scientific Acros
201825000 |
500 g | Plastic bottle |
64.62 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10474735
|
Thermo Scientific Acros
201820025 |
2.5 kg | Plastic bottle |
231.15 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
7783-28-0 | |
132.06 | |
MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
24540 | |
phosphoric acid diamine |
H9N2O4P | |
MFCD00010891 | |
diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
CHEBI:63051 | |
N.N.OP(O)(O)=O |
Specifications
7783-28-0 | |
7.9 to 8.3 (5% soln. at 20°C) | |
99+% | |
(NH4)2HPO4 | |
15, 540 | |
Solubility in water: 58g/100mL (20°C). Other solubilities: practically insoluble in alcohol and acetone | |
N.N.OP(O)(O)=O | |
132.06 | |
CHEBI:63051 | |
≥99% | |
1.61 |
White | |
Adhering Crystals | |
H9N2O4P | |
MFCD00010891 | |
diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
phosphoric acid diamine | |
24540 | |
132.06 | |
Analytical | |
Ammonium phosphate, dibasic, p.a. |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
missing translation for 'einecsNumber' : 231-987-8
missing translation for 'tsca' : TSCA
- Ammonium phosphate, dibasic
RUO – Research Use Only